2-Acetamido-3-brom-5-fluorpyridin

Lieferant: COMBI-BLOCKS

PY-7779-1G PY-7779-5G
COBBPY-7779-1GEA 0 EUR
COBBPY-7779-1G COBBPY-7779-5G
2-Acetamido-3-brom-5-fluorpyridin
2-Acetamido-3-brom-5-fluorpyridin
Formel: C₇H₆BrFN₂O
MDL: MFCD11053869
CAS-Nummer: 1065074-95-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR