2-Acetamido-5-amino-3-brompyridin
Lieferant: COMBI-BLOCKS
PY-7781-1G
PY-7781-250M
COBBPY-7781-1GEA
742
EUR
COBBPY-7781-1G
COBBPY-7781-250M
2-Acetamido-5-amino-3-brompyridin
2-Acetamido-5-amino-3-brompyridin
Formel:
C₇H₈BrN₃O |
MDL:
MFCD11053878 CAS-Nummer: 896161-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...