2-(Boc-amino)pyridin
Lieferant: COMBI-BLOCKS
Synonyme:
tert-Butyl N-(2-pyridyl)carbamate, tert-Butyl pyridin-2-ylcarbamate
PY-7791-1G
PY-7791-25G
PY-7791-5G
COBBPY-7791-1GEA
26
EUR
COBBPY-7791-1G
COBBPY-7791-25G
COBBPY-7791-5G
2-(Boc-amino)pyridin
2-(Boc-amino)pyridin
Formel:
C₁₀H₁₄N₂O₂ Molecular Weight: 194,23 g/mol |
MDL:
MFCD03411622 CAS-Nummer: 38427-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...