2-(Boc-amino)-6-chlorpyridin

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl (6-chloropyridin-2-yl)carbamate, 2-Amino-6-chloropyridine, 2-BOC protected

PY-7829-1G PY-7829-5G
COBBPY-7829-1GEA 102 EUR
COBBPY-7829-1G COBBPY-7829-5G
2-(Boc-amino)-6-chlorpyridin
2-(Boc-amino)-6-chlorpyridin
Formel: C₁₀H₁₃ClN₂O₂
Molecular Weight: 228,68 g/mol
MDL: MFCD09030821
CAS-Nummer: 159603-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR