2-Acetamido-6-brompyridin

Lieferant: COMBI-BLOCKS

Synonyme: N-(6-Bromopyridin-2-yl)acetamide

PY-7846-1G PY-7846-5G
COBBPY-7846-1GEA 127 EUR
COBBPY-7846-1G COBBPY-7846-5G
2-Acetamido-6-brompyridin
2-Acetamido-6-brompyridin
Formel: C₇H₇BrN₂O
Molecular Weight: 215,05 g/mol
MDL: MFCD00234277
CAS-Nummer: 25218-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR