2-Acetamido-5-brompyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Acetylamino-5-bromopyridine, N-(5-Bromopyridin-2-yl)acetamide

PY-7949-1G PY-7949-5G
COBBPY-7949-1GEA 26.2 EUR
COBBPY-7949-1G COBBPY-7949-5G
2-Acetamido-5-brompyridin
2-Acetamido-5-brompyridin
Formel: C₇H₇BrN₂O
Molecular Weight: 215,05 g/mol
MDL: MFCD00468968
CAS-Nummer: 7169-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR