3-Hydroxy-2,4,6-triiodbenzoesäure
Lieferant: FLUOROCHEM
Synonyme:
HTBA
001444-5G
001444-25G
FLUO001444-5GEA
72.9
EUR
FLUO001444-5G
FLUO001444-25G
3-Hydroxy-2,4,6-triiodbenzoesäure
3-Hydroxy-2,4,6-triiodobenzoic acid
Formel:
C₇H₃I₃O₃ Molecular Weight: 515,81 g/mol |
MDL:
MFCD00020305 CAS-Nummer: 53279-72-4 EINECS: 258-457-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...