2,2,2-Trifluorethyltoluol-4-sulfonat
Lieferant: FLUOROCHEM
Synonyme:
2,2,2-Trifluoroethyl p-toluenesulphonate, 2,2,2-Trifluoroethyl 4-methylbenzenesulphonate
001538-25G
001538-100G
001538-500G
FLUO001538-25GEA
34.1
EUR
FLUO001538-25G
FLUO001538-100G
FLUO001538-500G
2,2,2-Trifluorethyltoluol-4-sulfonat
2,2,2-Trifluorethyltoluol-4-sulfonat
Formel:
C₉H₉F₃O₃S Molecular Weight: 254.23 g/mol |
MDL:
MFCD00000443 CAS-Nummer: 433-06-7 EINECS: 207-085-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...