Bis(2-ethoxyethyl)adipat

Lieferant: FLUOROCHEM

001724-5G 001724-25G
FLUO001724-5GEA 61.1 EUR
FLUO001724-5G FLUO001724-25G
Bis(2-ethoxyethyl)adipat
Bis(2-ethoxyethyl)adipat
Formel: C₁₄H₂₆O₆
Molecular Weight: 290.36 g/mol
MDL: MFCD00041926
CAS-Nummer: 109-44-4
EINECS: 203-673-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR