1-(7-Methoxy-1-benzofuran-2-yl)ethanon
Lieferant: FLUOROCHEM
Synonyme:
1-(7-Methoxy-1-benzofuran-2-yl)-1-ethanone, 1-(7-Methoxybenzofuran-2-yl)ethanone
002487-1G
002487-5G
FLUO002487-1GEA
36.9
EUR
FLUO002487-1G
FLUO002487-5G
1-(7-Methoxy-1-benzofuran-2-yl)ethanon
2-Acetyl-7-methoxybenzofuran
Formel:
C₁₁H₁₀O₃ Molecular Weight: 190,2 g/mol |
MDL:
MFCD00016911 CAS-Nummer: 43071-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...