2,6-Difluor-3-nitrobenzonitril
Lieferant: FLUOROCHEM
004846-5G
004846-25G
FLUO004846-5GEA
33.8
EUR
FLUO004846-5G
FLUO004846-25G
2,6-Difluor-3-nitrobenzonitril
2,6-Difluor-3-nitrobenzonitril
Formel:
C₇H₂F₂N₂O₂ Molecular Weight: 184.1 g/mol |
MDL:
MFCD00042254 CAS-Nummer: 143879-77-0 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...