Ethyl-2,4-difluorbenzoat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2,4-difluorobenzoate

006098-5G 006098-25G
FLUO006098-5GEA 0 EUR
FLUO006098-5G FLUO006098-25G
Ethyl-2,4-difluorbenzoat
Ethyl-2,4-difluorbenzoat
Formel: C₉H₈F₂O₂
Molecular Weight: 186,16 g/mol
MDL: MFCD00153149
CAS-Nummer: 108928-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR