Ethyl-2-(trifluormethyl)benzoat
Lieferant: FLUOROCHEM
006145-1G
006145-5G
006145-25G
FLUO006145-1GEA
31.1
EUR
FLUO006145-1G
FLUO006145-5G
FLUO006145-25G
Ethyl-2-(trifluormethyl)benzoat
Ethyl-2-(trifluormethyl)benzoat
Formel:
C₁₀H₉F₃O₂ Molecular Weight: 218,18 g/mol |
MDL:
MFCD00013556 CAS-Nummer: 577-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...