Ethyl-4-(trifluormethyl)benzoat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 4-(trifluoromethyl)benzoate
006147-5G
006147-25G
006147-100G
006147-1G
FLUO006147-5GEA
39.9
EUR
FLUO006147-5G
FLUO006147-25G
FLUO006147-100G
FLUO006147-1G
Ethyl-4-(trifluormethyl)benzoat
Ethyl-4-(trifluormethyl)benzoat
Formel:
C₁₀H₉F₃O₂ Molecular Weight: 218,18 g/mol |
MDL:
MFCD00013563 CAS-Nummer: 583-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...