Indium(III)trifluormethansulfonat

Lieferant: FLUOROCHEM

Synonyme: Indium(III) trifluoromethanesulphonate, Indium triflate

006639-100G 006639-25G 006639-5G
FLUO006639-100GEA 0 EUR
FLUO006639-100G FLUO006639-25G FLUO006639-5G
Indium(III)trifluormethansulfonat
Indium(III)trifluormethansulfonat
Formel: C₃F₉InO₉S₃
Molecular Weight: 562.03 g/mol
MDL: MFCD00144478
CAS-Nummer: 128008-30-0
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR