Ethyl-3,4-difluorbenzoat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 3,4-difluorobenzoate
008303-5G
008303-25G
FLUO008303-5GEA
0
EUR
FLUO008303-5G
FLUO008303-25G
Ethyl-3,4-difluorbenzoat
Ethyl-3,4-difluorbenzoat
Formel:
C₉H₈F₂O₂ Molecular Weight: 186,16 g/mol |
MDL:
MFCD00272138 CAS-Nummer: 144267-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...