Methyl-2,4-difluorbenzoat
Lieferant: FLUOROCHEM
Synonyme:
Methyl 2,4-difluorobenzoate
008809-1G
008809-5G
008809-25G
FLUO008809-1GEA
31.1
EUR
FLUO008809-1G
FLUO008809-5G
FLUO008809-25G
Methyl-2,4-difluorbenzoat
Methyl-2,4-difluorbenzoat
Formel:
C₈H₆F₂O₂ Molecular Weight: 172,13 g/mol |
MDL:
MFCD03093799 CAS-Nummer: 106614-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...