Methyl-2,5-difluorbenzoat

Lieferant: FLUOROCHEM

Synonyme: Methyl 2,5-difluorobenzoate

008810-1G 008810-5G 008810-25G
FLUO008810-1GEA 31.1 EUR
FLUO008810-1G FLUO008810-5G FLUO008810-25G
Methyl-2,5-difluorbenzoat
Methyl-2,5-difluorbenzoat
Formel: C₈H₆F₂O₂
Molecular Weight: 172,13 g/mol
MDL: MFCD03093800
CAS-Nummer: 362601-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR