4-Bromphthalsäureanhydrid
Lieferant: FLUOROCHEM
Synonyme:
5-Bromoisobenzofuran-1,3-dione
009065-25G
009065-500G
FLUO009065-25GEA
55.2
EUR
FLUO009065-25G
FLUO009065-500G
4-Bromphthalsäureanhydrid
4-Bromphthalsäureanhydrid
Formel:
C₈H₃BrO₃ Molecular Weight: 227.01 g/mol |
MDL:
MFCD00191323 CAS-Nummer: 86-90-8 EINECS: 201-707-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...