Dimethyl-2-fluorterephthalat
Lieferant: FLUOROCHEM
Synonyme:
Dimethyl 2-fluoroterephthalate
009154-1G
009154-5G
FLUO009154-1GEA
121
EUR
FLUO009154-1G
FLUO009154-5G
Dimethyl-2-fluorterephthalat
Dimethyl-2-fluorterephthalat
Formel:
C₁₀H₉FO₄ Molecular Weight: 212,18 g/mol |
MDL:
MFCD00042266 CAS-Nummer: 5292-47-7 EINECS: 226-135-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...