2-Furohydrazid
Lieferant: CHEMPUR
Synonyme:
2-Furoic acid hydrazide, 2-Furoylhydrazine, 2-Furancarboxylic acid Hydrazide, 2-Furoic hydrazide, 2-Furoic acid Hydrazide, 2-Furancarboxylic acid hydrazide, 2-Furoic Hydrazide
FL9944.POR
009944-25G
CHMPFL9944.POREA
0
EUR
CHMPFL9944.POR
FLUO009944-25G
2-Furohydrazid
2-Furohydrazid
Formel:
C₅H₆N₂O₂ Molecular Weight: 126.11 g/mol |
MDL:
MFCD00003235 CAS-Nummer: 3326-71-4 EINECS: 222-046-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...