Lithiumbis(trifluormethylsulfonyl)imid

Lieferant: FLUOROCHEM

Synonyme: Lithium bis(trifluoromethanesulphonimide), Lithium bis((trifluoromethyl)sulphonyl)amide, Lithium Bis(Trifluoromethanesulphonyl)Imide, Bis(trifluoromethane)sulphonimide lithium salt

010326-25G 010326-100G
FLUO010326-25GEA 54.8 EUR
FLUO010326-25G FLUO010326-100G
Lithiumbis(trifluormethylsulfonyl)imid
Lithiumbis(trifluormethylsulfonyl)imid
Formel: C₂F₆LiNO₄S₂
Molecular Weight: 287.09 g/mol
MDL: MFCD00210017
CAS-Nummer: 90076-65-6
EINECS: 415-300-0
UN: 2923
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR