2-Amino-3,5-dibrom-6-methylpyridin
Lieferant: FLUOROCHEM
Synonyme:
3,5-Dibromo-6-methyl-2-pyridinamine
CFF292.25
010400-5G
010400-25G
CHMPCFF292.25EA
532
EUR
CHMPCFF292.25
FLUO010400-5G
FLUO010400-25G
2-Amino-3,5-dibrom-6-methylpyridin
2-Amino-3,5-dibromo-6-methylpyridine
Formel:
C₆H₆Br₂N₂ Molecular Weight: 265,94 g/mol |
MDL:
MFCD00068229 CAS-Nummer: 91872-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...