4-Chlorphthalsäureanhydrid

Lieferant: CHEMPUR

Synonyme: 5-Chloroisobenzofuran-1,3-dione

FL13485.POR 013485-1G
CHMPFL13485.POREA 0 EUR
CHMPFL13485.POR FLUO013485-1G
4-Chlorphthalsäureanhydrid
4-Chlorphthalsäureanhydrid
Formel: C₈H₃ClO₃
Molecular Weight: 182,56 g/mol
MDL: MFCD00152354
CAS-Nummer: 118-45-6
EINECS: 204-251-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR