5-Methyl-2-furoylchlorid

Lieferant: CHEMPUR

Synonyme: 5-Methyl-furan-2-carbonyl chloride

FL13648.POR 013648-2.5G 013648-1G
CHMPFL13648.POREA 0 EUR
CHMPFL13648.POR FLUO013648-2.5G FLUO013648-1G
5-Methyl-2-furoylchlorid
5-Methyl-2-furoylchlorid
Formel: C₆H₅ClO₂
Molecular Weight: 144,56 g/mol
MDL: MFCD03421416
CAS-Nummer: 14003-11-3
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR