3-Methyl-2-furancarbonsäure
Lieferant: CHEMPUR
Synonyme:
3-Methylfuran-2-carboxylic acid
FL13650.POR
013650-5G
013650-1G
CHMPFL13650.POREA
0
EUR
CHMPFL13650.POR
FLUO013650-5G
FLUO013650-1G
3-Methyl-2-furancarbonsäure
3-Methyl-2-furoic acid
Formel:
C₆H₆O₃ Molecular Weight: 126,11 g/mol |
MDL:
MFCD00014111 CAS-Nummer: 4412-96-8 EINECS: 224-571-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...