Methyl-3-methyl-2-furoat
Lieferant: CHEMPUR
Synonyme:
Methyl 3-methylfuran-2-carboxylate
FL13654.POR
013654-5G
013654-25G
CHMPFL13654.POREA
0
EUR
CHMPFL13654.POR
FLUO013654-5G
FLUO013654-25G
Methyl-3-methyl-2-furoat
Methyl-3-methyl-2-furoat
Formel:
C₇H₈O₃ Molecular Weight: 140.14 g/mol |
MDL:
MFCD00014110 CAS-Nummer: 6141-57-7 EINECS: 228-131-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...