Methyl-5-methyl-2-furoat
Lieferant: CHEMPUR
Synonyme:
Methyl 5-methylfuran-2-carboxylate
FL13655.POR
013655-5G
013655-1G
CHMPFL13655.POREA
0
EUR
CHMPFL13655.POR
FLUO013655-5G
FLUO013655-1G
Methyl-5-methyl-2-furoat
Methyl-5-methyl-2-furoat
Formel:
C₇H₈O₃ Molecular Weight: 140,14 g/mol |
MDL:
MFCD04116374 CAS-Nummer: 2527-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...