1-(2-Chlorbenzyl)piperazin
Lieferant: FLUOROCHEM
014883-1G
014883-5G
FLUO014883-1GEA
0
EUR
FLUO014883-1G
FLUO014883-5G
1-(2-Chlorbenzyl)piperazin
1-(2-Chlorbenzyl)piperazin
Formel:
C₁₁H₁₅ClN₂ Molecular Weight: 210.71 g/mol |
MDL:
MFCD01860843 CAS-Nummer: 17532-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...