1-(2,5-Dimethylbenzyl)piperazin

Lieferant: FLUOROCHEM

014890-100MG 014890-1G
FLUO014890-100MGEA 0 EUR
FLUO014890-100MG FLUO014890-1G
1-(2,5-Dimethylbenzyl)piperazin
1-(2,5-Dimethylbenzyl)piperazin
Formel: C₁₃H₂₀N₂
CAS-Nummer: 104481-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR