2-Methyl-5-phenyl-3-furancarbonsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Methyl-5-phenylfuran-3-carboxylic acid

CFF204.5 CFF204.10 017268-5G 017268-1G
CHMPCFF204.5EA 348 EUR
CHMPCFF204.5 CHMPCFF204.10 FLUO017268-5G FLUO017268-1G
2-Methyl-5-phenyl-3-furancarbonsäure
2-Methyl-5-phenyl-3-furoic acid
Formel: C₁₂H₁₀O₃
Molecular Weight: 202.21 g/mol
MDL: MFCD00221070
CAS-Nummer: 108124-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR