2-Acetamido-5-aminopyridin

Lieferant: FLUOROCHEM

Synonyme: 2-(Acetylamino)-5-aminopyridine

018015-1G 018015-5G
FLUO018015-1GEA 368 EUR
FLUO018015-1G FLUO018015-5G
2-Acetamido-5-aminopyridin
2-Acetamido-5-aminopyridin
Formel: C₇H₉N₃O
Molecular Weight: 151,17 g/mol
MDL: MFCD00160311
CAS-Nummer: 29958-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR