1-(4-(Trifluormethyl)phenyl)piperazin

Lieferant: FLUOROCHEM

Synonyme: N-(α,α,α-Trifluoro-p-tolyl)piperazine

018140-1G 018140-5G
FLUO018140-1GEA 0 EUR
FLUO018140-1G FLUO018140-5G
1-(4-(Trifluormethyl)phenyl)piperazin
1-(4-(Trifluormethyl)phenyl)piperazin
Formel: C₁₁H₁₃F₃N₂
Molecular Weight: 230,23 g/mol
MDL: MFCD00040765
CAS-Nummer: 30459-17-7
EINECS: 250-210-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR