1-(3,4-Dimethylbenzyl)piperazin

Lieferant: FLUOROCHEM

021567-5G 021567-25G
FLUO021567-5GEA 164 EUR
FLUO021567-5G FLUO021567-25G
1-(3,4-Dimethylbenzyl)piperazin
1-(3,4-Dimethylbenzyl)piperazin
Formel: C₁₃H₂₀N₂
CAS-Nummer: 212393-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR