1-(2-Brombenzyl)piperazin

Lieferant: FLUOROCHEM

021568-5G 021568-1G
FLUO021568-5GEA 0 EUR
FLUO021568-5G FLUO021568-1G
1-(2-Brombenzyl)piperazin
1-(2-Brombenzyl)piperazin
Formel: C₁₁H₁₅BrN₂
Molecular Weight: 255.16 g/mol
CAS-Nummer: 298705-59-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR