1-(3-Brombenzyl)piperazin

Lieferant: FLUOROCHEM

021569-5G 021569-1G
FLUO021569-5GEA 1100 EUR
FLUO021569-5G FLUO021569-1G
1-(3-Brombenzyl)piperazin
1-(3-Brombenzyl)piperazin
Formel: C₁₁H₁₅BrN₂
MDL: MFCD02211309
CAS-Nummer: 423154-81-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR