Ethyl-2,3-difluorbenzoat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 2,3-difluorobenzoate
023867-25G
023867-5G
FLUO023867-25GEA
480
EUR
FLUO023867-25G
FLUO023867-5G
Ethyl-2,3-difluorbenzoat
Ethyl-2,3-difluorbenzoat
Formel:
C₉H₈F₂O₂ Molecular Weight: 186,16 g/mol |
MDL:
MFCD06204337 CAS-Nummer: 773134-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...