Pyridinium-p-toluolsulfonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Pyridin-1-ium 4-methylbenzenesulphonate
M-Clarity™ Qualitätsstufe = MQ200
8.14330.0025
8.14330.0100
8.14330.0025EA
59.7
EUR
8.14330.0025
8.14330.0100
Pyridinium-p-toluolsulfonat, Sigma-Aldrich®
Pyridinium-p-toluolsulfonat
Formel:
C₁₂H₁₃NO₃S Molecular Weight: 251,31 g/mol |
MDL:
MFCD00013108 CAS-Nummer: 24057-28-1 EINECS: 246-002-7 |
Spezifikation Testergebnisse
Solubility | (20 °C) soluble |
Melting Point | 112 - 119 °C (dry substance) |
Molar Mass | 251.30 g/mol |
Hill Formula | C12H13NO3S |
Chemical Formula | 4-(CH3)C6H4SO3H * C5H5N |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...