Bis(2-ethylhexyl)adipat, Supelco®
Lieferant: Merck
|
Achtung
|
Synonyme:
Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate
M-Clarity™ Qualitätsstufe = MQ100
02138-1ML
02138-25ML
02138-5ML
02138-1MLEA
28.2
EUR
02138-1ML
02138-25ML
02138-5ML
Bis(2-ethylhexyl)adipat, Supelco®
Bis(2-ethylhexyl)adipat
Formel:
C₂₂H₄₂O₄ Molecular Weight: 370.57 g/mol |
MDL:
MFCD00009496 CAS-Nummer: 103-23-1 EINECS: 203-090-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...