2-Chlorethyltoluol-4-sulfonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Chloroethyl p-toluenesulphonate
M-Clarity™ Qualitätsstufe = MQ100
107999-100G
107999-25G
SIAL107999-100GEA
172
EUR
SIAL107999-100G
SIAL107999-25G
2-Chlorethyltoluol-4-sulfonat, Sigma-Aldrich®
2-Chlorethyltoluol-4-sulfonat
Formel:
C₉H₁₁ClO₃S Molecular Weight: 234,7 g/mol |
MDL:
MFCD00000970 CAS-Nummer: 80-41-1 EINECS: 201-277-2 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...