Saccharin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
o-Sulphobenzimide, o-Benzoic sulphimide
M-Clarity™ Qualitätsstufe = MQ100
109185-250G
109185-500G
SIAL109185-250GEA
94.3
EUR
SIAL109185-250G
SIAL109185-500G
Saccharin, Sigma-Aldrich®
Saccharin
Formel:
C₇H₅NO₃S Molecular Weight: 183,19 g/mol |
MDL:
MFCD00005866 CAS-Nummer: 81-07-2 EINECS: 201-321-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...