Ethyl-3-furoat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Ethyl furan-3-carboxylate, Ethyl 3-Furoate, Ethyl 3-Furancarboxylate, 3-Furancarboxylic acid ethyl ester, 3-Furoic acid ethyl ester
M-Clarity™ Qualitätsstufe = MQ100
164542-25G
164542-5G
SIAL164542-25GEA
372
EUR
SIAL164542-25G
SIAL164542-5G
Ethyl-3-furoat, Sigma-Aldrich®
Ethyl-3-furoat
Formel:
C₇H₈O₃ Molecular Weight: 140,14 g/mol |
MDL:
MFCD00005348 CAS-Nummer: 614-98-2 EINECS: 210-403-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...