Triethyloxoniumhexafluorophosphat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

164682-25G 164682-5G
SIAL164682-25GEA 567 EUR
SIAL164682-25G SIAL164682-5G
Triethyloxoniumhexafluorophosphat, Sigma-Aldrich®
Triethyloxonium hexafluorophosphate
Formel: C₆H₁₅F₆OP
Molecular Weight: 189,99 g/mol
CAS-Nummer: 17950-40-2
EINECS: 241-876-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR