Saccharin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: o-Sulphobenzimide, o-Benzoic sulphimide

M-Clarity™ Qualitätsstufe = MQ200

240931-1G 240931-50G
SIAL240931-1GEA 45.3 EUR
SIAL240931-1G SIAL240931-50G
Saccharin, Sigma-Aldrich®
Saccharin
Formel: C₇H₅NO₃S
Molecular Weight: 183,19 g/mol
MDL: MFCD00005866
CAS-Nummer: 81-07-2
EINECS: 201-321-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR