Methyl-3-brompropionat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 3-bromopropanoate

M-Clarity™ Qualitätsstufe = MQ200

242373-100G 242373-25G 242373-5G
SIAL242373-100GEA 181 EUR
SIAL242373-100G SIAL242373-25G SIAL242373-5G
Methyl-3-brompropionat, Sigma-Aldrich®
Methyl-3-brompropionat
Formel: C₄H₇BrO₂
Molecular Weight: 167 g/mol
MDL: MFCD00000250
CAS-Nummer: 3395-91-3
EINECS: 222-247-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR