(Triphenylphosphin)gold(I)chlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Chlorotriphenylphosphine gold(I), Chloro(triphenylphosphine)gold(I), Triphenylphosphinechlorogold
M-Clarity™ Qualitätsstufe = MQ100
254037-500MG
254037-5G
SIAL254037-500MGEA
192
EUR
SIAL254037-500MG
SIAL254037-5G
(Triphenylphosphin)gold(I)chlorid, Sigma-Aldrich®
(Triphenylphosphine)gold(I) chloride
Formel:
C₁₈H₁₅AuClP Molecular Weight: 494,71 g/mol |
MDL:
MFCD00009588 CAS-Nummer: 14243-64-2 EINECS: 238-117-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...