Bis(triphenylphosphin)nickel(II)bromid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Nickel(II) bromide bis(triphenylphosphine), Dibromobis(triphenylphosphine)nickel(II)
M-Clarity™ Qualitätsstufe = MQ100
331708-10G
331708-50G
SIAL331708-10GEA
108
EUR
SIAL331708-10G
SIAL331708-50G
Bis(triphenylphosphin)nickel(II)bromid, Sigma-Aldrich®
Bis(triphenylphosphin)nickel(II)bromid
Formel:
C₃₆H₃₀Br₂NiP₂ Molecular Weight: 743,08 g/mol |
MDL:
MFCD00015865 CAS-Nummer: 14126-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...