Chlor(diisopropyl)phosphin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Di-i-propylchlorophosphine
M-Clarity™ Qualitätsstufe = MQ100
337773-25G
337773-5G
SIAL337773-25GEA
275
EUR
SIAL337773-25G
SIAL337773-5G
Chlor(diisopropyl)phosphin, Sigma-Aldrich®
Chlor(diisopropyl)phosphin
Formel:
C₆H₁₄ClP Molecular Weight: 152,6 g/mol |
MDL:
MFCD00015027 CAS-Nummer: 40244-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...