Methyl-3-methylcrotonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 3,3-dimethylacrylate, Methyl-3,3-dimethylacrylate

M-Clarity™ Qualitätsstufe = MQ100

368571-25G 368571-5G
SIAL368571-25GEA 236 EUR
SIAL368571-25G SIAL368571-5G
Methyl-3-methylcrotonat, Sigma-Aldrich®
Methyl-3-methylcrotonat
Formel: C₆H₁₀O₂
Molecular Weight: 114.14 g/mol
MDL: MFCD00043940
CAS-Nummer: 924-50-5
EINECS: 213-107-4
UN: 3272
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR