Zinn(II)trifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tin trifluoromethanesulphonate, Tin triflate, Tin(II) trifluoromethanesulphonate

M-Clarity™ Qualitätsstufe = MQ200

388122-1G 388122-5G
SIAL388122-1GEA 97.5 EUR
SIAL388122-1G SIAL388122-5G
Zinn(II)trifluormethansulfonat, Sigma-Aldrich®
Zinn(II)trifluormethansulfonat
Formel: C₂F₆O₆S₂Sn
Molecular Weight: 416,85 g/mol
MDL: MFCD00191251
CAS-Nummer: 62086-04-8
UN: 3288
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR